Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ 2,4,5-Trimethylaniline Solution, 100 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111020017 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0017 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0017 Supplier MilliporeSigma Supelco Supplier No. 310392ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,4,5-Trimethylaniline is a primary aromatic amine (PAA) commonly used in chemical industries. PAAs are persistent environmental water pollutants and known to be potential human carcinogens.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 137-17-7
Density 0.78 g/mL
Flash Point -5°C
Linear Formula C9H13 N
MDL Number MFCD00025201
Type Analytical Standard
Quantity 2 mL
Formula Weight 135.21
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 μg/mL in acetonitrile
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.