Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ 2,4-D Methyl Ester Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022519 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2519 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2519 Supplier MilliporeSigma Supelco Supplier No. 45416100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 1928-38-7
Physical Form Neat
Molecular Formula C9H8Cl2O3
Linear Formula C9H8Cl2O3
MDL Number MFCD00018356
UN Number UN 3077 9 / PGIII
Synonym Methyl (2,4-dichlorophenoxy)acetate
Quantity 100 mg
Formula Weight 235.06
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.