Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ 2,4-DDT Solution, 100 μg/mL in Methanol, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111017317 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7317 2 mL
11-101-7163 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-7317 Supplier MilliporeSigma Supelco Supplier No. 458392ML
Add to Cart
Add to Cart

Specifications

Product Line PESTANAL
Concentration 100 μg/mL in methanol
Linear Formula C14H9Cl5
MDL Number MFCD00036120
Synonym 1,1,1-Trichloro-2-(2-chlorophenyl)-2-(4-chlorophenyl)ethane solution
Quantity 2 mL
Formula Weight 354.49
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.