Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ 2-Aminopyridine, 98-100% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020576 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0576 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0576 Supplier MilliporeSigma Supelco Supplier No. 366851G
Add to Cart
Add to Cart

2-Aminopyridine can be called as structural motifs, which can be encountered in a variety of useful compounds, that includes fluorescent organic materials and therapeutics. It can undergo condensation with an aromatic aldehyde to give the corresponding schiff′s base.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 504-29-0
Flash Point 92°C - Closed Cup
Physical Form Neat
Linear Formula C5H6N2
MDL Number MFCD00006312
Melting Point 54°C to 58°C
Boiling Point 204°C to 210°C
Synonym 2-AP; 2-Pyridinamine; 2-Pyridylamine
Type Analytical Standard
Quantity 1 g
Formula Weight 94.11
Percent Purity 98 to 100% (GC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.