Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ 3-Methylcholanthrene Solution, 100 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111020060 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0060 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0060 Supplier MilliporeSigma Supelco Supplier No. 4643410MLR
Add to Cart
Add to Cart

3-Methylcholanthrene is a potent carcinogenic polycyclic aromatic hydrocarbon. It plays a role in the pathogenesis of human colon cancer.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 56-49-5
Flash Point 2°C
Linear Formula C21H16
MDL Number MFCD00003704
Type Analytical Standard
Quantity 10 mL
Formula Weight 268.35
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 μg/mL in acetonitrile

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.