Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ 4-Aminophenol, MilliporeSigma™ Supelco™

Catalog No. 111020666 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0666 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0666 Supplier MilliporeSigma Supelco Supplier No. 358371G
Add to Cart
Add to Cart

4-Aminophenol can be obtained via the reduction of 4-nitrophenol with excess of sodium borohydride. It can also be obtained as a primary hydrolytic degradation product of paracetamol.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 123-30-8
Flash Point 195°C - Closed Cup
Physical Form Neat
Linear Formula H2 NC6H4 OH
MDL Number MFCD00007869
Melting Point 185°C to 189°C
Synonym 4-Hydroxyaniline
Solubility Slightly soluble in water
Type Analytical Standard
Quantity 1 g
Formula Weight 109.13
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.