Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ 4-Chloroaniline, MilliporeSigma™ Supelco™

Catalog No. 111020650 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0650 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0650 Supplier MilliporeSigma Supelco Supplier No. 358231G
Add to Cart
Add to Cart

4-Chloroaniline is a chlorinated aromatic amines, which can generally find applications in industry in the production of cosmetics, dyes, herbicides and pharmaceutical products. It is also a major metabolite of the phenyl-urea herbicides.

Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 106-47-8
Vapor Pressure 0.15 mmHg (25°C)
Vapor Density 4.4 (vs. air)
Flash Point 120.0°C - Closed Cup
Physical Form Neat
Linear Formula ClC6H4 NH2
MDL Number MFCD00007835
Melting Point 67°C to 70°C
Boiling Point 232°C
Type Analytical Standard
Quantity 1 g
Formula Weight 127.57
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.