Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ 4-Nitrophenol, 100% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111020504 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0504 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0504 Supplier MilliporeSigma Supelco Supplier No. 358361G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
Ignition Point 541°F
CAS 100-02-7
Vapor Pressure 0.6 mmHg (120°C)
Flash Point 169°C
Physical Form Neat
Linear Formula O2 NC6H4 OH
MDL Number MFCD00007331
Melting Point 110°C to 115°C
Boiling Point 279°C
Synonym p-Nitrophenol
Type Analytical Standard
Quantity 1 g
Formula Weight 139.11
Percent Purity 100% (HPLC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.