Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ 5-Hydroxythiabendazole, MilliporeSigma™ Supelco™

Catalog No. 111018241 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8241 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8241 Supplier MilliporeSigma Supelco Supplier No. 3381810MG
Add to Cart
Add to Cart

5-Hydroxythiabendazole present in excreta, eggs, edible tissues and milk is found to be the major metabolite of thiabendazole, an active ingredient in fungicidal preparations effective for plants. It can also be used as an anthelmintic drug, which is applicable both for humans and animals.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 948-71-0
Physical Form Neat
Linear Formula C10H7N3 OS
MDL Number MFCD00152219
Synonym 2-(4-Thiazolyl)-5-benzimidazolol; 5-Hydroxy-2-(4-thiazolyl)benzimidazole
Type Analytical Standard
Quantity 10 mg
Formula Weight 217.25
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.