Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Aclonifen, MilliporeSigma™ Supelco™

Catalog No. 111019438 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9438 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9438 Supplier MilliporeSigma Supelco Supplier No. 3679250MGR
Add to Cart
Add to Cart

Aclonifen is a diphenyl-ether herbicide, which can be used for weed control management, in specific for sunflowers. Its mode of action involves, the inhibition of carotenoid synthesis and suppression of protoporphyrinogen oxidase enzyme involved in the chlorophyll synthesis pathway.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 74070-46-5
Physical Form Neat
Linear Formula C12H9 ClN2O3
MDL Number MFCD00143542
Synonym 2-Chloro-6-nitro-3-phenoxyaniline
Type Analytical Standard
Quantity 50 mg
Formula Weight 264.66
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.