Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Alachlor OA, MilliporeSigma™ Supelco™

Catalog No. 111013332 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3332 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3332 Supplier MilliporeSigma Supelco Supplier No. 3414610MG
Add to Cart
Add to Cart

Alachlor OA is a metabolite of alachlor, a preemergence herbicide used for the control of annual grasses and many broad-leaved weeds in corn and other crops.

Agriculture and Environmental
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 171262-17-2
Physical Form Neat
Linear Formula C14H19 NO4
MDL Number MFCD08702642
Synonym N-(2,6-Diethylphenyl)-N-(methoxymethyl)oxalamic acid; [(2,6-Diethylphenyl)(methoxymethyl)amino]oxo-acetic acid
Type Analytical Standard
Quantity 10 mg
Formula Weight 265.30
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.