Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Aldicarb, MilliporeSigma™ Supelco™

Catalog No. 111019528 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9528 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9528 Supplier MilliporeSigma Supelco Supplier No. 33386100MG
Add to Cart
Add to Cart

Aldicarb is a white crystalline, N-methycarbamate ester of aliphatic oxime, which may be used as an insecticide in agriculture.

Agriculture and Environmental
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 116-06-3
Flash Point >100°C
Physical Form Neat
Linear Formula C7H14N2O2 S
MDL Number MFCD00041807
Synonym O-(Methylcarbamoyl)-2-methyl-2-(methylthio)propionaldehyd-oxime
Type Analytical Standard
Quantity 100 mg
Formula Weight 190.26
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.