Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Aldrin, MilliporeSigma™ Supelco™

Catalog No. 111019377 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9377 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9377 Supplier MilliporeSigma Supelco Supplier No. 3666625MG
Add to Cart
Add to Cart

Aldrin is a chlorinated cyclodiene, which is a synthetically produced pyrethroid insecticide.

Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 309-00-2
Physical Form Neat
Linear Formula C12H8 Cl6
MDL Number MFCD00135587
Type Analytical Standard
Quantity 25 mg
Formula Weight 364.91
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.