Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Allyl isothiocyanate, 99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020067 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0067 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0067 Supplier MilliporeSigma Supelco Supplier No. 366821G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Allyl isothiocyanate is a naturally occurring compound in plants, and antibacterial agent, which belongs to the class of cruciferae. It can also serve as a promising candidate that can show anti-cancer activity in both cultured cells and animal models.

Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottled Under Inert Gas
Product Line PESTANAL
CAS 57-06-7
Density 1.013 g/mL (at 25°C)
Flash Point 46°C - Closed Cup
Physical Form Neat
Linear Formula CH2=CHCH2 NCS
MDL Number MFCD00004822
Melting Point -80°C
Boiling Point 150°C
Synonym AITC; Oil of mustard
Type Analytical Standard
Quantity 1 g
Formula Weight 99.15
Percent Purity 99.7% (GC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.