Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Amidosulfuron, MilliporeSigma™ Supelco™

Catalog No. 111012755 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2755 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2755 Supplier MilliporeSigma Supelco Supplier No. 33588100MG
Add to Cart
Add to Cart

Amidosulfuron is a selective, sulfonylurea herbicide employed for the control of many grasses and broadleaf weeds in crop protection of vines, rice, citrus, corn, potatoes and tomatoes

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 120923-37-7
Physical Form Neat
Linear Formula C9H15N5O7S2
MDL Number MFCD01310639
Synonym 1-(4,6-Dimethoxypyrimidin-2-yl)-3-(N-mesyl-N-methylsulfamoyl)urea
Type Analytical Standard
Quantity 100 mg
Formula Weight 369.37
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.