Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Amisulbrom, MilliporeSigma™ Supelco™

Catalog No. 111018834 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8834 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8834 Supplier MilliporeSigma Supelco Supplier No. 3284925MG
Add to Cart
Add to Cart

Amisulbrom is classified under the triazole group of fungicides. It contains a sulfone functional group with potent bioactivity.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 348635-87-0
Physical Form Neat
Linear Formula C13H13 BrFN5O4S2
MDL Number MFCD11865361
Synonym 3-(3-Bromo-6-fluoro-2-methylindol-1-ylsulfonyl)-N,N-dimethyl-1 H-1,2,4-triazole-1-sulfonamide
Type Analytical Standard
Quantity 25 mg
Formula Weight 466.31
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.