Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Atrazine-desethyl, MilliporeSigma™ Supelco™

Catalog No. 111014912 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4912 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4912 Supplier MilliporeSigma Supelco Supplier No. 36629250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706023

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 6190-65-4
Physical Form Neat
Linear Formula C6H10 ClN5
MDL Number MFCD00078646
Synonym 2-Amino-4-chloro-6-isopropylamino-1,3,5-triazine; Desethylatrazine
Type Analytical Standard
Quantity 250 mg
Formula Weight 187.63
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.