Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Atrazine, MilliporeSigma™ Supelco™

Catalog No. 111020160 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0160 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0160 Supplier MilliporeSigma Supelco Supplier No. 45330250MGR
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706023

Agriculture and Environmental
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00041810
Synonym 2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine

Specifications

CAS 1912-24-9
Ignition Point 779°F
Linear Formula C8H14 ClN5
Quantity 250 mg
Vapor Density 3.2 (vs. air)
Vapor Pressure 13.8 mmHg (21.1°C)
Formula Weight 215.68
Grade Puriss.
Physical Form Neat
Product Line PESTANAL
ShelfLife Limited shelf life, expiry date on the label
RTECSNumber XY5600000

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.