Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Beflubutamid Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022456 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2456 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2456 Supplier MilliporeSigma Supelco Supplier No. 32866100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 113614-08-7
Physical Form Neat
Molecular Formula C18H17F4NO2
Linear Formula C18H17F4NO2
MDL Number MFCD04112601
UN Number UN 3077 9 / PGIII
Synonym 2-[4-Fluoro-3-(trifluoromethyl)phenoxy]-N-(phenylmethyl)butanamide; N-Benzyl-2-(alpha,alpha,alpha,4-tetrafluoro-m-tolyloxy)butyramide
Quantity 100 mg
Formula Weight 355.33
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.