Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Benalaxyl, MilliporeSigma™ Supelco™

Catalog No. 111019708 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9708 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9708 Supplier MilliporeSigma Supelco Supplier No. 31222250MG
Add to Cart
Add to Cart

Benalaxyl is a systemic fungicide belonging to the acylalanine family. It is found to be active against Oomycetes belonging to Peronosporaceae family.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 71626-11-4
Physical Form Neat
Linear Formula C20H23 NO3
MDL Number MFCD00143652
Type Analytical Standard
Quantity 250 mg
Formula Weight 325.40
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.