Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Benthiavalicarb isopropyl, MilliporeSigma™ Supelco™

Catalog No. 111013366 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3366 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3366 Supplier MilliporeSigma Supelco Supplier No. 3300610MG
Add to Cart
Add to Cart

Benthiavalicarb isopropyl is classified under the class of amino acid amide carbamate category of fungicides.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 177406-68-7
Physical Form Neat
Linear Formula C18H24 FN3O3 S
MDL Number MFCD08690472
Type Analytical Standard
Quantity 10 mg
Formula Weight 381.46
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.