Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ β-BHC Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022578 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2578 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2578 Supplier MilliporeSigma Supelco Supplier No. 48494
Add to Cart
Add to Cart

β-BHC belongs to the class of organochlorine compounds

TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Physical Form Neat
Molecular Formula C6H6Cl6
Linear Formula C6H6Cl6
MDL Number MFCD00135948
UN Number UN2811 - Class 6.1 - PG 3 - EHS - RQ - Toxic solids, organic, n.o.s. HI: all
Quantity 50 mg
Formula Weight 290.83
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.