Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Bromocyclen, MilliporeSigma™ Supelco™

Catalog No. 111013333 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3333 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3333 Supplier MilliporeSigma Supelco Supplier No. 33398100MG
Add to Cart
Add to Cart

Bromocyclen is a cyclodiene insecticide and has been used widely against ectoparasites for treating domestic animals.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 1715-40-8
Physical Form Neat
Linear Formula C8H5 BrCl6
MDL Number MFCD00151378
Synonym 5- Bromomethyl-1,2,3,4,7,7-hexachloro-2-norbornene
Type Analytical Standard
Quantity 100 mg
Formula Weight 393.75
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.