Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Buprofezin-(isopropyl-1,1,1,3,3,3-d6), Analytical Standard, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111001603 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-1603 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-1603 Supplier MilliporeSigma Supelco Supplier No. 915025MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Product Line PESTANAL
Linear Formula C16D6H17N3OS
Melting Point 104°C to 108°C
Synonym Buprofezin-d6
Quantity 5 mg
Formula Weight 311.48
Percent Purity ≥98.0% (GC)
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.