Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Cadusafos, MilliporeSigma™ Supelco™

Catalog No. 111018875 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8875 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8875 Supplier MilliporeSigma Supelco Supplier No. 3250550MG
Add to Cart
Add to Cart

Cadusafos is a nematicide used in controlling nematodes, soil insects and cutworms in a variety of fruits and vegetables. Its mode of action involves the inhibition of acetylcholinesterase.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 95465-99-9
Flash Point 129.4°C
Physical Form Neat
Linear Formula C10H23O2 PS2
MDL Number MFCD00072511
Synonym O-Ethyl S,S-bis(1-methylpropyl) phosphorodithioate
Type Analytical Standard
Quantity 50 mg
Formula Weight 270.39
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.