Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Carbofuran, MilliporeSigma™ Supelco™

Catalog No. 111020498 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0498 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0498 Supplier MilliporeSigma Supelco Supplier No. 32056250MG
Add to Cart
Add to Cart

Carbofuran belongs to N-methylcarbamate class of chemicals used to control soil-dwelling and foliar-feeding insects on agricultural crops. Carbofuran acts by inhibiting cholinesterase enzyme activity.

Agriculture and Environmental
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 1563-66-2
Physical Form Neat
Linear Formula C12H15 NO3
MDL Number MFCD00041819
Melting Point 150°C to 153°C
Synonym 2,3-Dihydro-2,2-dimethyl-7-benzofuranol N-methylcarbamate
Type Analytical Standard
Quantity 250 mg
Formula Weight 221.25
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.