Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Carpropamid, MilliporeSigma™ Supelco™

Catalog No. 111019187 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9187 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9187 Supplier MilliporeSigma Supelco Supplier No. 31682100MG
Add to Cart
Add to Cart

Carpropamid is widely used for the control of rice blast caused by Pyricularia oryzae. Carpropamid acts by inhibiting scytalone dehydratase enzyme involved in fungal melanin biosynthesis in a phytopathogenic fungus.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 104030-54-8
Physical Form Neat
Linear Formula C15H18 Cl3 NO
MDL Number MFCD03095718
Synonym 2,2-Dichloro-N-[1-(4-chlorophenyl)ethyl]-1-ethyl-3-methylcyclopropanecarboxamide
Type Analytical Standard
Quantity 100 mg
Formula Weight 334.67
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.