Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Chloroacetic acid, 98-100% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111020188 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0188 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0188 Supplier MilliporeSigma Supelco Supplier No. 365441G
Add to Cart
Add to Cart

Chloroacetic acid is the degradation product of the herbicide, chloroacetanilide.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 79-11-8
Vapor Pressure 0.75 mmHg (20°C)
Vapor Density 3.26 (vs. air)
Physical Form Neat
Linear Formula ClCH2 COOH
MDL Number MFCD00002683
Melting Point 60°C to 63°C
Boiling Point 189°C
Synonym Monochloroacetic acid
Type Analytical Standard
Quantity 1 g
Formula Weight 94.50
Percent Purity 98 to 100% (HPLC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.