Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Chlorpyrifos, MilliporeSigma™ Supelco™

Catalog No. 111019515 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9515 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9515 Supplier MilliporeSigma Supelco Supplier No. 45395100MG
Add to Cart
Add to Cart

Chlorpyrifos belongs to the organophosphorus class of insecticides, effectively used for the control of a variety of insect pests such as mosquitoes, fire ants, cockroaches, horn flies, lice, fleas, turf and ornamental plant insects.

Agriculture and Environmental
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 2921-88-2
Physical Form Neat
Linear Formula C9H11 Cl3 NO3 PS
MDL Number MFCD00041800
Synonym Chlorpyriphos
Type Analytical Standard
Quantity 100 mg
Formula Weight 350.59
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.