Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Clethodim, Mixture of isomers, MilliporeSigma™ Supelco™

Catalog No. 111016014 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6014 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6014 Supplier MilliporeSigma Supelco Supplier No. 3419050MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clethodim is a cyclohexanedione herbicide, which inhibits the activity of acetyl-coenzyme A carboxylase. It is widely used to control annual and perennial grass weeds of broad leaved crops, vegetables, vines and trees.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 99129-21-2
Physical Form Neat
Linear Formula C17H26 ClNO3 S
MDL Number MFCD01632327
Synonym 2-{(E)-1-[(E)-3-Chloroallyloxyimino]propyl}-5-[2-(ethylthio)propyl]-3-hydroxy-2-cyclohexen-1-one
Type Analytical Standard
Quantity 50 mg
Formula Weight 359.91
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.