Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Clofentezine, MilliporeSigma™ Supelco™

Catalog No. 111018965 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8965 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8965 Supplier MilliporeSigma Supelco Supplier No. 3676350MG
Add to Cart
Add to Cart
Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 74115-24-5
Physical Form Neat
Linear Formula C14H8 Cl2N4
MDL Number MFCD00143998
Melting Point 182°C to 185°C
Synonym 3,6-Bis(2-chlorophenyl)-1,2,4,5-tetrazine; Clofentezine
Type Analytical Standard
Quantity 50 mg
Formula Weight 303.15
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.