Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Cyanophos, MilliporeSigma™ Supelco™

Catalog No. 111018868 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8868 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8868 Supplier MilliporeSigma Supelco Supplier No. 4627925MG
Add to Cart
Add to Cart

Cyanophos is an organophosphate insecticide, which is effectively used against various root-parasitic nematodes such as Diaspididae, Lepidoptera, Coccidae, Aphididae and other insects on various fruits and vegetables.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 2636-26-2
Flash Point >100°C
Physical Form Neat
Linear Formula C9H10 NO3 PS
MDL Number MFCD00055485
Synonym O-(4-Cyanophenyl) O,O-dimethyl phosphorothioate
Type Analytical Standard
Quantity 25 mg
Formula Weight 243.22
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.