Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Cyflumetofen, MilliporeSigma™ Supelco™

Catalog No. 111018604 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8604 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8604 Supplier MilliporeSigma Supelco Supplier No. 3206850MG
Add to Cart
Add to Cart

Cyflumetofen is an acaricide, which facilitates the rapid knockdown of spider mites and phytophagous mites. Its mode of action involves the inhibition of mitochondrial electron transport and is suitable for use in integrated pest management (IPM) systems.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 400882-07-7
Physical Form Neat
Linear Formula C24H24F3 NO4
MDL Number MFCD11112207
Synonym 2-Methoxyethyl (RS)-2-(4-tert-butylphenyl)-2-cyano-3-oxo-3-(alpha,alpha,alpha-trifluoro-o-tolyl)propionate
Type Analytical Standard
Quantity 50 mg
Formula Weight 447.45
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.