Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Cypermethrin Solution, 100 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111018904 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8904 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8904 Supplier MilliporeSigma Supelco Supplier No. 4583510ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Agriculture and Environmental
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 52315-07-8
Flash Point 2°C
Linear Formula C22H19 Cl2 NO3
MDL Number MFCD00055328
Type Analytical Standard
Quantity 10 mL
Formula Weight 416.30
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 μg/mL in acetonitrile

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.