Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Cyproconazol, MilliporeSigma™ Supelco™

Catalog No. 111018883 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8883 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8883 Supplier MilliporeSigma Supelco Supplier No. 46068100MG
Add to Cart
Add to Cart

Cyproconazol is an azole compound fungicide, commonly used as a wood preservative and as a biocide for protecting fruits and vegetable crops.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 94361-06-5
Flash Point >100°C
Physical Form Neat
Linear Formula C15H18 ClN3 O
MDL Number MFCD01678672
Type Analytical Standard
Quantity 100 mg
Formula Weight 291.78
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.