Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Cyprosulfamide, MilliporeSigma™ Supelco™

Catalog No. 111013569 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3569 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3569 Supplier MilliporeSigma Supelco Supplier No. 32522100MG
Add to Cart
Add to Cart

Cyprosulfamide is a herbicide, and can protect the plants from salinity stress and induced vigorous growth, including the formation of new tillers and early flowering.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 221667-31-8
Physical Form Neat
Linear Formula C18H18N2O5 S
MDL Number MFCD15146679
Synonym N-[4-(Cyclopropylaminocarbonyl)phenylsulfonyl]-2-methoxybenzamide
Type Analytical Standard
Quantity 100 mg
Formula Weight 374.41
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.