Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Cyromazin, MilliporeSigma™ Supelco™

Catalog No. 111015109 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5109 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5109 Supplier MilliporeSigma Supelco Supplier No. 4541450MG
Add to Cart
Add to Cart
Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 66215-27-8
Flash Point 100°C
Physical Form Neat
Linear Formula C6H10N6
MDL Number MFCD00078650
Melting Point 223°C to 227°C
Synonym N-Cyclopropyl-1,3,5-triazin-2,4,6-triamine; Cyromazin
Type Analytical Standard
Quantity 50 mg
Formula Weight 166.18
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.