Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Cyromazine-(cyclopropyl-2,2,3,3-d4), ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012789 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2789 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2789 Supplier MilliporeSigma Supelco Supplier No. 931015MG
Add to Cart
Add to Cart

Specifications

Product Line PESTANAL
CAS 1219804-19-9
Linear Formula C6D4H6N6
MDL Number MFCD11616807
Synonym N-(Cyclopropyl-2,2,3,3-d4)-1,3,5-triazin-2,4,6-triamine; Cyromazin-d4; Cyromazine-d4
Type Analytical Standard
Quantity 5 mg
Formula Weight 170.21
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.