Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Daminozide, MilliporeSigma™ Supelco™

Catalog No. 111013273 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3273 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3273 Supplier MilliporeSigma Supelco Supplier No. 45418250MG
Add to Cart
Add to Cart

Daminozide is a commonly used plant growth regulator, which acts by inhibiting 2-oxoglutarate-dependent dioxygenase activity in the gibberellin biosynthesis pathway.

Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 1596-84-5
Physical Form Neat
Linear Formula HO2 CCH2 CH2 CONHN(CH3)2
MDL Number MFCD00002787
Melting Point 162°C to 164°C
Synonym Succinic acid mono(2,2-dimethylhydrazide)
Type Analytical Standard
Quantity 250 mg
Formula Weight 160.17
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.