Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Demeton-S-methyl Solution, 100 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111019222 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9222 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9222 Supplier MilliporeSigma Supelco Supplier No. 342342ML
Add to Cart
Add to Cart

Demeton-S-methyl is a metabolite of the pesticide thiometon, used for food, environmental, and toxicology analyses.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 919-86-8
Flash Point 2°C
Linear Formula C6H15O3 PS2
MDL Number MFCD00055464
Synonym S-[2-(Ethylthio)ethyl] O,O-dimethyl phosphorothioate; Methyldemetone
Type Analytical Standard
Quantity 2 mL
Formula Weight 230.29
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 μg/mL in acetonitrile
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.