Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Dibutyl phthalate, ≥98%, MilliporeSigma™ Supelco™

Catalog No. 111020543 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0543 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0543 Supplier MilliporeSigma Supelco Supplier No. 367361G
Add to Cart
Add to Cart

Dibutyl phthalate can be widely used as a plasticizer and additive in special paints and adhesives. It can find applications as an endocrine disrupter and peroxisome proliferator, which can act against demodicidosis infection.

Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
Ignition Point 756°F
CAS 84-74-2
Vapor Pressure 1 mmHg (147°C)
Vapor Density 9.6 (vs. air)
Density 1.043 g/mL (at 25°C)
Flash Point 171°C
Physical Form Neat
Linear Formula C6H4-1,2-[CO2(CH2)3 CH3]2
MDL Number MFCD00009441
Melting Point -35°C
Boiling Point 340°C
Synonym n-Butyl phthalate; DBP; Phthalic acid dibutyl ester
Type Analytical Standard
Quantity 1 g
Formula Weight 278.34
Percent Purity ≥98%
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.