Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Dichlon, MilliporeSigma™ Supelco™

Catalog No. 111020605 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0605 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0605 Supplier MilliporeSigma Supelco Supplier No. 454341EA
Add to Cart
Add to Cart

Dichlon is an agricultural fungicide of the quinone class, widely used against a variety of plant pathogenic fungi.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 117-80-6
Physical Form Neat
Linear Formula C10H4 Cl2O2
MDL Number MFCD00001677
Melting Point 194°C to 197°C
Synonym 2,3-Dichloro-1,4-naphthoquinone; Dichlon
Type Analytical Standard
Quantity 1 Ea.
Formula Weight 227.04
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.