Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ (±)-Diclofop, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014156 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4156 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4156 Supplier MilliporeSigma Supelco Supplier No. 5225625MG
Add to Cart
Add to Cart

Specifications

CAS 40843-25-2
Product Line PESTANAL
Physical Form Neat
Linear Formula C15H12 Cl2O4
MDL Number MFCD00152216
Synonym (RS)-2-[4-(2,4-Dichlorophenoxy)phenoxy]propionic acid
Type Analytical Standard
Quantity 25 mg
Formula Weight 327.16
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) carbon content, 54.0-56.2%
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.