Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Dieldrin, MilliporeSigma™ Supelco™

Catalog No. 111019920 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9920 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9920 Supplier MilliporeSigma Supelco Supplier No. 33491100MGR
Add to Cart
Add to Cart

Dieldrin is a chlorinated hydrocarbon insecticide, which is one of the persistent contaminants present in the environment.

Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 60-57-1
Vapor Density 13.2 (vs. air)
Physical Form Neat
Linear Formula C12H8 Cl6 O
MDL Number MFCD00135591
Melting Point 143°C to 144°C
Synonym 1,2,3,4,10,10-Hexachloro-1,4,4 A,5,6,7,8,8 A-octahydro-6,7-epoxy-1,4:5,8-dimethanonaphthalene
Type Analytical Standard
Quantity 100 mg
Formula Weight 380.91
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.