Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Dimethachlor Metabolite SYN 528702 sodium salt, MilliporeSigma™ Supelco™

Catalog No. 111016375 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6375 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6375 Supplier MilliporeSigma Supelco Supplier No. 3249725MG
Add to Cart
Add to Cart
Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 1228182-52-2
Physical Form Neat
Linear Formula C15H20 NNaO5 S
MDL Number MFCD16652582
Synonym Sodium 3-{2-[(2,6-dimethylphenyl)(2-hydroxyacetyl)amino]ethylsulfanyl}-2-hydroxypropionate
Type Analytical Standard
Quantity 25 mg
Formula Weight 349.38
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.