Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Dimethyl phthalate, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020446 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0446 1 mL
11-101-9110 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-0446 Supplier MilliporeSigma Supelco Supplier No. 413201MLF
Add to Cart
Add to Cart

Dimethyl phthalate can widely find applications in industries, for the manufacture of polyvinyl chloride and cellulose ester-based plastics like cellulose acetate and cellulose butyrate.

Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
Ignition Point 1033°F
CAS 131-11-3
Density 1.19 g/mL (at 25°C)
Flash Point 146°C
Physical Form Neat
Linear Formula C6H4-1,2-(CO2 CH3)2
MDL Number MFCD00008425
Melting Point 2°C
Boiling Point 282°C
Synonym DMP
Type Analytical Standard
Quantity 1 mL
Formula Weight 194.18
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.