Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Diphenylamine, MilliporeSigma™ Supelco™

Catalog No. 111020673 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0673 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0673 Supplier MilliporeSigma Supelco Supplier No. 45456250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diphenylamine is an organic compound, used for post-harvest deterioration of apple and pear crops. It finds applications in rubber production, dyes, polymers, pesticides, pharmaceutical, lubricating oils and photography chemicals.

Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottled Under Inert Gas
Product Line PESTANAL
Ignition Point 1175°F
CAS 122-39-4
Vapor Pressure 1 mmHg (108°C)
Vapor Density 5.82 (vs. air)
Flash Point 153°C
Physical Form Neat
Linear Formula (C6H5)2 NH
MDL Number MFCD00003014
Melting Point 50°C to 53°C
Boiling Point 302°C
Type Analytical Standard
Quantity 250 mg
Formula Weight 169.22
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.