Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Emamectin benzoate, MilliporeSigma™ Supelco™

Catalog No. 111018887 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8887 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8887 Supplier MilliporeSigma Supelco Supplier No. 31733250MG
Add to Cart
Add to Cart

Emamectin benzoate is a macrocyclic insecticide found to be stable aqueous hydrolysis. Avermectin B1 can be the starting material synthesizing emamectin benzoate. In some occasions, it is used as acaricide for vegetables, fruits and cereals.

TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 155569-91-8
Physical Form Neat
MDL Number MFCD01757274
Synonym (4 ′′R)-4 ′′-Deoxy-4 ′′-(methylamino)avermectin B1 benzoate
Type Analytical Standard
Quantity 250 mg
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.