Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Ethoxyquin, MilliporeSigma™ Supelco™

Catalog No. 111015901 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5901 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5901 Supplier MilliporeSigma Supelco Supplier No. 31519250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethoxyquin (EQ), an aromatic compound, is an antioxidant mostly used in animal and fish feeds. It is seen to inhibit carcinogenic effects of polycyclic aromatic hydrocarbons. Oxidation initiated by air, light or transition metals in unsaturated lipids can be stopped from propagation by EQ.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 91-53-2
Density 1.03 g/mL (at 20°C)
Shipping Condition Wet ice
Physical Form Neat
Linear Formula C14H19 NO
MDL Number MFCD00023883
Synonym 1,2-Dihydro-6-ethoxy-2,2,4-trimethylquinoline; 6-Ethoxy-1,2-dihydro-2,2,4-trimethylquinoline
Type Analytical Standard
Quantity 250 mg
Formula Weight 217.31
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.