Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Etridiazole, MilliporeSigma™ Supelco™

Catalog No. 111019193 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9193 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9193 Supplier MilliporeSigma Supelco Supplier No. 34340100MGR
Add to Cart
Add to Cart

Etridiazole is a fungicide, which is found to be effective against the oomycete group of fungi. It is used for seed treatment of various crops, in cotton cultivation during planting stage and non-bearing coffee and citrus fruits by horticultural nurseries.

Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 2593-15-9
Flash Point 154.5°C - open°Cup
Physical Form Neat
Linear Formula C5H5 Cl3N2 OS
MDL Number MFCD00071812
Synonym 5-Ethoxy-3-trichloromethyl-1,2,4-thiadiazole; Ethazol
Type Analytical Standard
Quantity 100 mg
Formula Weight 247.53
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.